Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Per3
Entrez Gene ID
18628
Gene Name
period circadian clock 3
Gene Aliases
2810049O06Rik, mPer3
Chromosome Location
Chr.4: 151003655 - 151044675 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289877.1 NP_001276806.1 7 - 8 1297 106
NM_001289878.1 NP_001276807.1 8 - 9 1346 106
NM_011067.3 NP_035197.2 7 - 8 1297 106
GenBank mRNA AB013605.1 - 7 - 8 1104 106
AF050182.1 - 7 - 8 1221 106
AJ007377.1 - 7 - 8 1104 106
AK036345.1 - 7 - 8 1214 106
AK166514.1 - 7 - 8 1298 106
BC141032.1 - 7 - 8 895 106
BC145178.1 - 8 - 9 944 106

View Full Product Details