Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnfrsf9
Entrez Gene ID
21942
Gene Name
tumor necrosis factor receptor superfamily, member 9
Gene Aliases
4-1BB, A930040I11Rik, AA408498, AI325004, CDw137, Cd137, ILA, Ly63
Chromosome Location
Chr.4: 150920155 - 150946104 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077508.1 NP_001070976.1 5 - 6 543 72
NM_001077509.1 NP_001070977.1 5 - 6 524 72
NM_011612.2 NP_035742.1 5 - 6 543 72
GenBank mRNA AK019885.1 - 5 - 6 510 72
AK154625.1 - 5 - 6 527 72
BC028507.1 - 5 - 6 480 72
DQ832278.1 - 4 - 5 429 72
DQ832279.1 - 4 - 5 429 72

View Full Product Details