The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025820.3 | NP_080096.1 | 13 - 14 | 1994 | 121 |
GenBank mRNA | AK004749.1 | - | 13 - 14 | 1995 | 121 |
AK012962.1 | - | 13 - 14 | 2005 | 121 | |
AK020002.1 | - | 13 - 14 | 2011 | 121 | |
AK088882.1 | - | 13 - 14 | 1998 | 121 | |
BC029187.1 | - | 13 - 14 | 1959 | 121 |