Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc30a4
Entrez Gene ID
22785
Gene Name
solute carrier family 30 (zinc transporter), member 4
Gene Aliases
Znt4, lm, znT-4
Chromosome Location
Chr.2: 122681233 - 122702663 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290993.1 NP_001277922.1 3 - 4 994 112
NM_011774.3 NP_035904.2 4 - 5 1141 112
GenBank mRNA AF003747.1 - 4 - 5 942 112
AK134245.1 - 4 - 5 1119 112
AK166106.1 - 3 - 4 996 112
BC117997.1 - 4 - 5 999 112
CV556160.1 - 2 - 3 305 112

View Full Product Details