Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dtwd1
Entrez Gene ID
69185
Gene Name
DTW domain containing 1
Gene Aliases
1810033A06Rik
Chromosome Location
Chr.2: 126152113 - 126165277 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026981.2 NP_081257.1 4 - 5 784 93
GenBank mRNA AK007678.1 - 3 - 4 751 93
AK149746.1 - 3 - 4 4334 93
BC020149.1 - 4 - 5 731 93

View Full Product Details