Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cyp39a1
- Entrez Gene ID
-
56050
- Gene Name
- cytochrome P450, family 39, subfamily a, polypeptide 1
- Gene Aliases
-
mCYP39A1
- Chromosome Location
-
Chr.17: 43667372 - 43751431 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC021418.1 |
- |
3 - 4 |
598 |
77 |
View Full Product Details