Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mut
Entrez Gene ID
17850
Gene Name
methylmalonyl-Coenzyme A mutase
Gene Aliases
D230010K02Rik, Mcm
Chromosome Location
Chr.17: 40934685 - 40961989 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008650.3 NP_032676.2 1 - 2 94 66
GenBank mRNA AK146309.1 - 1 - 2 54 66
BY262299.1 - 1 - 2 95 66

View Full Product Details