The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290991.1 | NP_001277920.1 | 12 - 13 | 1970 | 153 |
NM_001290992.1 | NP_001277921.1 | 12 - 13 | 1969 | 153 | |
NM_212450.4 | NP_997615.1 | 12 - 13 | 1973 | 153 | |
GenBank mRNA | AK028251.1 | - | 10 - 11 | 1033 | 153 |
AK030085.1 | - | 12 - 13 | 1489 | 153 | |
AK033382.1 | - | 12 - 13 | 1522 | 153 | |
AK044805.1 | - | 12 - 13 | 1504 | 153 | |
AK049463.1 | - | 12 - 13 | 1489 | 153 | |
AK053513.1 | - | 12 - 13 | 1499 | 153 | |
AK082646.1 | - | 12 - 13 | 1829 | 153 | |
AK163262.1 | - | 12 - 13 | 1974 | 153 | |
BC052660.1 | - | 12 - 13 | 1456 | 153 | |
BC066219.1 | - | 12 - 13 | 1490 | 153 |