Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fermt1
Entrez Gene ID
241639
Gene Name
fermitin family homolog 1 (Drosophila)
Gene Aliases
5830467P10Rik, Kindlin-1
Chromosome Location
Chr.2: 132904177 - 132946036 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198029.2 NP_932146.2 7 - 8 1689 67
GenBank mRNA AF073489.1 - 4 - 5 475 67
AK050804.1 - 6 - 7 837 67

View Full Product Details