Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ntsr2
Entrez Gene ID
18217
Gene Name
neurotensin receptor 2
Gene Aliases
NT2R, NTRL
Chromosome Location
Chr.12: 16653452 - 16660236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008747.2 NP_032773.2 1 - 2 655 106
GenBank mRNA AB041826.1 - 1 - 2 631 106
AB041827.1 - 1 - 2 631 106
AK005368.1 - 1 - 2 370 106
AK081073.1 - 1 - 2 388 106
AK158943.1 - 1 - 2 655 106
BC141019.1 - 1 - 2 650 106

View Full Product Details