Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ntsr2
Entrez Gene ID
18217
Gene Name
neurotensin receptor 2
Gene Aliases
NT2R, NTRL
Chromosome Location
Chr.12: 16653452 - 16660236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008747.2 NP_032773.2 3 - 4 1017 92
GenBank mRNA AB041826.1 - 3 - 4 993 92
AB041827.1 - 3 - 4 993 92
AK005368.1 - 3 - 4 674 92
AK081073.1 - 2 - 3 2245 92
AK158943.1 - 3 - 4 1017 92
BC141019.1 - 3 - 4 1012 92
U51908.1 - 3 - 4 1016 92

View Full Product Details