The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163818.1 | NP_001157290.1 | 5 - 6 | 913 | 61 |
GenBank mRNA | AJ504718.1 | - | 5 - 6 | 919 | 61 |
AK053183.1 | - | 6 - 7 | 932 | 61 | |
AK145485.1 | - | 4 - 5 | 357 | 61 | |
BC030903.1 | - | 5 - 6 | 920 | 61 | |
BC052059.1 | - | 5 - 6 | 916 | 61 |