Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plin4
Entrez Gene ID
57435
Gene Name
perilipin 4
Gene Aliases
S3-12, mKIAA1881
Chromosome Location
Chr.17: 56100587 - 56109947 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020568.3 NP_065593.2 7 - 8 3928 74

View Full Product Details