The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020568.3 | NP_065593.2 | 8 - 9 | 4020 | 109 |
GenBank mRNA | AF064748.1 | - | 8 - 9 | 4031 | 109 |
AK080156.1 | - | 2 - 3 | 109 | 109 | |
AK173286.1 | - | 8 - 9 | 2748 | 109 |