The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159523.1 | NP_001152995.1 | 1 - 2 | 224 | 69 |
NM_009168.2 | NP_033194.1 | 1 - 2 | 224 | 69 | |
GenBank mRNA | AB018423.1 | - | 1 - 2 | 223 | 69 |
AK142491.1 | - | 1 - 2 | 240 | 69 | |
AK164639.1 | - | 1 - 2 | 160 | 69 | |
BC103798.1 | - | 1 - 2 | 117 | 69 |