The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159523.1 | NP_001152995.1 | 6 - 7 | 1077 | 63 |
NM_009168.2 | NP_033194.1 | 6 - 7 | 1074 | 63 | |
GenBank mRNA | AB018423.1 | - | 6 - 7 | 1073 | 63 |
AK142491.1 | - | 6 - 7 | 1090 | 63 | |
AK164639.1 | - | 6 - 7 | 1010 | 63 | |
BC052026.1 | - | 5 - 6 | 2004 | 63 | |
BC079574.1 | - | 5 - 6 | 2004 | 63 | |
BC103798.1 | - | 6 - 7 | 970 | 63 |