Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cstl1
Entrez Gene ID
228756
Gene Name
cystatin-like 1
Gene Aliases
RCET1, RCET2, RCET3
Chromosome Location
Chr.2: 148750361 - 148755378 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177655.3 NP_808323.1 3 - 4 499 77
GenBank mRNA AK131797.1 - 3 - 4 500 77
AK132807.1 - 3 - 4 502 77
AY706755.1 - 3 - 4 343 77
AY706756.1 - 3 - 4 343 77
AY707915.1 - 3 - 4 499 77
BC048646.1 - 3 - 4 499 77

View Full Product Details