Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uvrag
Entrez Gene ID
78610
Gene Name
UV radiation resistance associated gene
Gene Aliases
9530039D02Rik, AI648770, BB124205, Uvrag1, Uvragl
Chromosome Location
Chr.7: 98886742 - 99141168 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178635.3 NP_848750.3 13 - 14 1498 75
GenBank mRNA AK030814.1 - 13 - 14 1429 75
AK043084.1 - 13 - 14 1476 75
AK078085.1 - 13 - 14 1490 75
AK089900.1 - 13 - 14 1483 75
AK154903.1 - 13 - 14 1498 75
AK165185.1 - 13 - 14 1479 75
AK202347.1 - 2 - 3 256 75
BC034072.1 - 8 - 9 860 75
BC034176.1 - 13 - 14 1467 75

View Full Product Details