Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uvrag
Entrez Gene ID
78610
Gene Name
UV radiation resistance associated gene
Gene Aliases
9530039D02Rik, AI648770, BB124205, Uvrag1, Uvragl
Chromosome Location
Chr.7: 98886742 - 99141168 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178635.3 NP_848750.3 1 - 2 309 64
GenBank mRNA AK020590.1 - 1 - 2 284 64
AK030814.1 - 1 - 2 240 64
AK043084.1 - 1 - 2 287 64
AK044129.1 - 1 - 2 292 64
AK078085.1 - 1 - 2 301 64
AK089900.1 - 1 - 2 294 64
AK154903.1 - 1 - 2 309 64
AK165015.1 - 1 - 2 297 64
AK165185.1 - 1 - 2 289 64
BC034176.1 - 1 - 2 278 64

View Full Product Details