Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mocs1
Entrez Gene ID
56738
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
3110045D15Rik, AI043170, AW536397
Chromosome Location
Chr.17: 49428364 - 49455435 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020042.2 NP_064426.2 3 - 4 502 100
NM_028464.1 NP_082740.1 3 - 4 502 100
GenBank mRNA AK157231.1 - 3 - 4 458 100
BC049914.1 - 1 - 2 130 100
CN701969.1 - 3 - 4 471 100

View Full Product Details