Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Daam2
Entrez Gene ID
76441
Gene Name
dishevelled associated activator of morphogenesis 2
Gene Aliases
2310016D11Rik, AI843643, AW557870
Chromosome Location
Chr.17: 49456022 - 49564491 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008231.2 NP_001008232.2 23 - 24 3020 83
GenBank mRNA AK009382.1 - 8 - 9 842 83
AK122266.1 - 21 - 22 5938 83
AY426536.1 - 23 - 24 3049 83
BC037720.1 - 7 - 8 741 83

View Full Product Details