Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Unc5cl
- Entrez Gene ID
-
76589
- Gene Name
- unc-5 homolog C (C. elegans)-like
- Gene Aliases
-
2510009H09Rik, BB155270, Zud
- Chromosome Location
-
Chr.17: 48453839 - 48539709 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK039924.1 |
- |
4 - 5 |
395 |
61 |
View Full Product Details