Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alg8
Entrez Gene ID
381903
Gene Name
asparagine-linked glycosylation 8 (alpha-1,3-glucosyltransferase)
Gene Aliases
AI447372, Gm1089
Chromosome Location
Chr.7: 97371594 - 97392159 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199035.2 NP_950200.2 8 - 9 922 110
GenBank mRNA AK141341.1 - 8 - 9 919 110
AK166755.1 - 8 - 9 917 110
AK166897.1 - 8 - 9 923 110
BC061244.1 - 8 - 9 911 110

View Full Product Details