The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020024.3 | NP_064408.2 | 4 - 5 | 589 | 91 |
GenBank mRNA | AK047115.1 | - | - | 591 | 91 |
AK139383.1 | - | - | 599 | 91 | |
BC031418.1 | - | 4 - 5 | 589 | 91 | |
BC080311.1 | - | 4 - 5 | 590 | 91 |