Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slx1b
Entrez Gene ID
75764
Gene Name
SLX1 structure-specific endonuclease subunit homolog B (S. cerevisiae)
Gene Aliases
1110030E23Rik, 2410170E21Rik, 4833422P03Rik, AI853643, Giyd1, Giyd2
Chromosome Location
Chr.7: 126688927 - 126695783 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029420.4 NP_083696.2 5 - 6 969 68
NR_033446.1 4 - 5 917 68
GenBank mRNA AK010748.1 - 5 - 6 969 68
AK010828.1 - 4 - 5 1038 68
AK049139.1 - 5 - 6 970 68
AK053901.1 - 4 - 5 858 68
BC055909.1 - 5 - 6 920 68
BC145468.1 - 4 - 5 781 68
BC145688.1 - 5 - 6 834 68

View Full Product Details