Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spns1
Entrez Gene ID
73658
Gene Name
spinster homolog 1
Gene Aliases
2210013K02Rik, Spin1, Spinl
Chromosome Location
Chr.7: 126370060 - 126377934 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023712.3 NP_076201.2 1 - 2 1097 85
NR_045537.1 1 - 2 1097 85
GenBank mRNA AF212372.1 - 1 - 2 588 85
AK008721.1 - 1 - 2 594 85
AK033907.1 - 1 - 2 574 85
AK154353.1 - 1 - 2 587 85
AK166931.1 - 1 - 2 589 85
BC002297.1 - 1 - 2 116 85
BC026854.1 - 1 - 2 558 85
BC085491.1 - 1 - 2 541 85

View Full Product Details