The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023712.3 | NP_076201.2 | 4 - 5 | 1449 | 218 |
NR_045537.1 | 5 - 6 | 1428 | 197 | ||
GenBank mRNA | AF212372.1 | - | 4 - 5 | 940 | 218 |
AK008721.1 | - | 5 - 6 | 925 | 197 | |
AK154353.1 | - | 4 - 5 | 939 | 218 | |
AK166931.1 | - | 4 - 5 | 941 | 218 | |
BC002297.1 | - | 4 - 5 | 468 | 218 | |
BC026854.1 | - | 4 - 5 | 910 | 218 | |
BC085491.1 | - | 4 - 5 | 893 | 218 |