Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ckmt1
- Entrez Gene ID
-
12716
- Gene Name
- creatine kinase, mitochondrial 1, ubiquitous
- Gene Aliases
-
Mt-CK, ScCKmit, UbCKmit, mi-CK
- Chromosome Location
-
Chr.2: 121357751 - 121363737 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC025976.1 |
- |
1 - 2 |
69 |
69 |
View Full Product Details