Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prokr2
Entrez Gene ID
246313
Gene Name
prokineticin receptor 2
Gene Aliases
B830005M06Rik, EG-VEGRF2, Gpcr73l1, Gpr73l1, PKR2
Chromosome Location
Chr.2: 132370329 - 132385498 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BB844646.1 - 1 - 2 194 57

View Full Product Details