The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008027.2 | NP_032053.1 | 12 - 13 | 1412 | 56 |
GenBank mRNA | AK150692.1 | - | 12 - 13 | 1434 | 56 |
AK150827.1 | - | 12 - 13 | 1361 | 56 | |
AK167323.1 | - | 12 - 13 | 1339 | 56 | |
AK200487.1 | - | 1 - 2 | 40 | 56 |