Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nsmce4a
Entrez Gene ID
67872
Gene Name
non-SMC element 4 homolog A (S. cerevisiae)
Gene Aliases
2410003A14Rik
Chromosome Location
Chr.7: 130532526 - 130547381 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162855.1 NP_001156327.1 1 - 2 388 62
GenBank mRNA AK010349.1 - 1 - 2 389 62
AK010795.1 - 1 - 2 354 62
BC032990.1 - 10 - 11 62
BC051572.1 - 10 - 11 62
BE951712.1 - 1 - 2 265 62

View Full Product Details