Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx5
Entrez Gene ID
69178
Gene Name
sorting nexin 5
Gene Aliases
0910001N05Rik, 1810032P22Rik, AU019504, D2Ertd52e
Chromosome Location
Chr.2: 144250123 - 144270902 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199188.1 NP_001186117.1 6 - 7 741 120
NM_024225.5 NP_077187.1 5 - 6 652 120
GenBank mRNA AK003091.1 - - 365 120
AK007676.1 - 5 - 6 600 120
AK029463.1 - 5 - 6 612 120
AK031873.1 - 6 - 7 633 120
AK049129.1 - 6 - 7 633 120
AK078776.1 - 6 - 7 634 120
AK152311.1 - 6 - 7 677 121
AK159115.1 - 2 - 3 195 120
AK159904.1 - 5 - 6 557 120
AK167362.1 - 5 - 6 572 120
AK169786.1 - 6 - 7 633 120
BC002242.1 - 6 - 7 710 120

View Full Product Details