Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agbl2
Entrez Gene ID
271813
Gene Name
ATP/GTP binding protein-like 2
Gene Aliases
4930524K04, A430081C19Rik, CCP2
Chromosome Location
Chr.2: 90782587 - 90834437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178755.3 NP_848870.2 4 - 5 391 73
GenBank mRNA AK029744.1 - 4 - 5 424 73
AK029932.1 - 4 - 5 392 73
AK030138.1 - 4 - 5 412 73
AK030182.1 - 4 - 5 438 73
AK040259.1 - 4 - 5 327 73
AK079691.1 - 4 - 5 328 73
DQ867026.1 - 4 - 5 424 73
DQ867027.1 - 4 - 5 424 73
DQ867028.1 - 4 - 5 424 73
DQ867029.1 - 4 - 5 424 73

View Full Product Details