Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pamr1
Entrez Gene ID
210622
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
5930437L24, E430002G05Rik, Ramp
Chromosome Location
Chr.2: 102550021 - 102643041 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173749.4 NP_776110.3 1 - 2 241 65
GenBank mRNA AK031254.1 - 1 - 2 114 65
AK088017.1 - 1 - 2 140 65
BB620582.1 - 1 - 2 108 65
BC031841.1 - 1 - 2 195 65
BC057685.1 - 1 - 2 195 65

View Full Product Details