Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spred1
Entrez Gene ID
114715
Gene Name
sprouty protein with EVH-1 domain 1, related sequence
Gene Aliases
5730461F13Rik, AL024345, AW047647
Chromosome Location
Chr.2: 117121071 - 117182279 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277256.1 NP_001264185.1 1 - 2 681 75
NM_033524.3 NP_277059.1 1 - 2 681 75
GenBank mRNA AK155065.1 - 1 - 2 644 75
AK155070.1 - 1 - 2 683 75
BC057874.1 - 1 - 2 114 75
BC079606.1 - 1 - 2 302 75

View Full Product Details