Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spred1
Entrez Gene ID
114715
Gene Name
sprouty protein with EVH-1 domain 1, related sequence
Gene Aliases
5730461F13Rik, AL024345, AW047647
Chromosome Location
Chr.2: 117121071 - 117182279 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277256.1 NP_001264185.1 2 - 3 864 98
NM_033524.3 NP_277059.1 2 - 3 864 98
GenBank mRNA AB063495.1 - 2 - 3 210 98
AK155065.1 - 2 - 3 827 98
AK155070.1 - 2 - 3 866 98
BC041685.1 - 1 - 2 178 98
BC057874.1 - 2 - 3 297 98
BC079606.1 - 2 - 3 485 98

View Full Product Details