The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039160.2 | NP_001034249.2 | 1 - 2 | 114 | 89 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243040.1 | NP_001229969.1 | 1 - 2 | 111 | 89 |
GenBank mRNA | AY167973.1 | - | 1 - 2 | 84 | 89 |