The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159548.1 | NP_001153020.1 | 1 - 2 | 119 | 69 |
NM_001159549.1 | NP_001153021.1 | 1 - 2 | 119 | 69 | |
GenBank mRNA | CN537360.1 | - | 1 - 2 | 95 | 69 |
CN537407.1 | - | 1 - 2 | 119 | 69 |