The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001097623.1 | NP_001091092.1 | 7 - 8 | 2121 | 96 |
NM_172622.2 | NP_766210.1 | 4 - 5 | 1958 | 96 | |
GenBank mRNA | AK035176.1 | - | 2 - 3 | 1728 | 96 |
AK046822.1 | - | 2 - 3 | 1599 | 96 | |
AK154542.1 | - | 6 - 7 | 2134 | 96 | |
AK155049.1 | - | 6 - 7 | 2235 | 96 | |
AK155404.1 | - | 6 - 7 | 2132 | 96 | |
BC059215.1 | - | 7 - 8 | 2121 | 96 | |
BC108416.1 | - | 2 - 3 | 1581 | 96 |