Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alx4
Entrez Gene ID
11695
Gene Name
aristaless-like homeobox 4
Gene Aliases
lst
Chromosome Location
Chr.2: 93642434 - 93681341 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007442.3 NP_031468.1 3 - 4 1090 96
GenBank mRNA AF001465.1 - 4 - 5 1060 96

View Full Product Details