Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pin1rt1
Entrez Gene ID
241593
Gene Name
protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1, retrogene 1
Gene Aliases
OTTMUSG00000014964, Pin1-ps1, Pin1l
Chromosome Location
Chr.2: 104713887 - 104716362 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033768.2 NP_001028940.1 1 - 2 212 61
GenBank mRNA AK145372.1 - 1 - 2 213 61
BC147362.1 - 1 - 2 34 61
BC147363.1 - 1 - 2 34 61

View Full Product Details