Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim72
Entrez Gene ID
434246
Gene Name
tripartite motif-containing 72
Gene Aliases
BC067209, MG53
Chromosome Location
Chr.7: 128003426 - 128011393 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079932.3 NP_001073401.1 2 - 3 591 98
GenBank mRNA AB231474.1 - 2 - 3 495 98
BC067209.1 - 2 - 3 157 98
BC147789.1 - 2 - 3 591 98
BC158111.1 - 2 - 3 576 98
BC158120.1 - 2 - 3 576 98
BC172156.1 - 2 - 3 591 98

View Full Product Details