Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf39
Entrez Gene ID
386454
Gene Name
ring finger protein 39
Gene Aliases
BC038661, Gm1786, Gm1787, Gm320, LIRF
Chromosome Location
Chr.17: 36943051 - 36947986 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099632.1 NP_001093102.1 1 - 2 401 66
GenBank mRNA BC038661.1 - 1 - 2 471 66
BC051578.1 - 1 - 2 480 66
BC132405.1 - 1 - 2 401 66
BC145677.1 - 1 - 2 401 66

View Full Product Details