The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025593.1 | NP_079869.1 | 1 - 2 | 126 | 64 |
GenBank mRNA | AK011021.1 | - | 1 - 2 | 106 | 64 |
AK149137.1 | - | 1 - 2 | 106 | 64 | |
AK168159.1 | - | 1 - 2 | 106 | 64 | |
BY124873.1 | - | 1 - 2 | 128 | 64 |