The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081446.1 | NP_001074915.1 | 8 - 9 | 805 | 70 |
GenBank mRNA | AK138400.1 | - | 7 - 8 | 741 | 70 |
AK160429.1 | - | 2 - 3 | 207 | 70 |