The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025846.2 | NP_080122.2 | 1 - 2 | 389 | 95 |
GenBank mRNA | AK011419.1 | - | 1 - 2 | 389 | 95 |
AK078870.1 | - | 1 - 2 | 390 | 95 | |
AK172037.1 | - | 1 - 2 | 381 | 95 | |
AK172113.1 | - | 1 - 2 | 375 | 95 | |
BC003871.1 | - | 1 - 2 | 340 | 95 |