The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164258.1 | NP_001157730.1 | 17 - 18 | 2189 | 90 |
GenBank mRNA | AK154558.1 | - | 18 - 19 | 2292 | 90 |
BC151014.1 | - | 17 - 18 | 2189 | 90 | |
BC151020.1 | - | 17 - 18 | 2189 | 90 |