The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164258.1 | NP_001157730.1 | 4 - 5 | 421 | 114 |
GenBank mRNA | AK154558.1 | - | 5 - 6 | 524 | 114 |
BC151014.1 | - | 4 - 5 | 421 | 114 | |
BC151020.1 | - | 4 - 5 | 421 | 114 |