Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crym
Entrez Gene ID
12971
Gene Name
crystallin, mu
Gene Aliases
-
Chromosome Location
Chr.7: 120186384 - 120201988 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016669.1 NP_057878.1 3 - 4 454 107
GenBank mRNA AF039391.1 - 3 - 4 454 107
AK138422.1 - 3 - 4 425 107
AK143116.1 - 3 - 4 444 107
AK163781.1 - 3 - 4 425 107
BC045159.1 - 3 - 4 415 107

View Full Product Details