Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddr2
Entrez Gene ID
18214
Gene Name
discoidin domain receptor family, member 2
Gene Aliases
AW495251, Ntrkr3, tyro10
Chromosome Location
Chr.1: 169972307 - 170110836 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022563.2 NP_072075.2 6 - 7 849 123
GenBank mRNA AK132504.1 - 3 - 4 347 123
BC138826.1 - 7 - 8 1095 123
BC138827.1 - 7 - 8 990 123
G49238.1 - 7 - 8 1156 123
X76505.1 - 7 - 8 1156 123

View Full Product Details